(2-Hydroxyethyl)triphenylphosphonium chloride 95%
Supplier: Ambeed
|
Warning
|
A835818-1G
A835818-5G
77146-822EA
6.37
USD
77146-822
77146-824
(2-Hydroxyethyl)triphenylphosphonium chloride 95%
(2-Hydroxyethyl)triphenylphosphonium chloride
Precaución: for Research Use Only
Formula:
C₂₀H₂₀ClOP MW: 342.8 g/mol |
MDL Number:
MFCD00011754 CAS Number: 23250-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...