(2-Chlorophenyl)(phenyl)methanol 95%

Supplier: Ambeed
Warning

A835352-100MG A835352-250MG A835352-1G
77146-818EA 28.65 USD
77146-818 77146-820 77686-862
(2-Chlorophenyl)(phenyl)methanol 95%
(2-Chlorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
MW: 218.67 g/mol
MDL Number: MFCD00030010
CAS Number: 6954-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR