2,1,3-Benzothiadiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Benzo[c][1,2,5]thiadiazole-5-carbaldehyde

A832529-100MG A832529-1G A832529-250MG
77146-804EA 41.2 USD
77146-804 77146-806 77146-808
2,1,3-Benzothiadiazole-5-carbaldehyde 97%
2,1,3-Benzothiadiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₄N₂OS
MDL Number: MFCD00457949
CAS Number: 71605-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR