1-Carboxy-N,N,N-trimethylmethanaminium hydroxide 97%

Supplier: Ambeed
Danger

A829571-1000G A829571-25G A829571-500G
77146-776EA 60.77 USD
77146-776 77146-778 77146-780
1-Carboxy-N,N,N-trimethylmethanaminium hydroxide 97%
1-Carboxy-N,N,N-trimethylmethanaminium hydroxide

Precaución: for Research Use Only

Formula: C₅H₁₃NO₃
MDL Number: MFCD00150010
CAS Number: 590-47-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR