1-Allyl-3-methylimidazolium chloride 98%

Supplier: Ambeed
Warning

A829438-5G A829438-100G
77146-774EA 7.21 USD
77146-774 77569-548
1-Allyl-3-methylimidazolium chloride 98%
1-Allyl-3-methylimidazolium chloride

Precaución: For research use only.

Formula: C₇H₁₁ClN₂
MDL Number: MFCD07784450
CAS Number: 65039-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR