(3,6-Dichloro-2-methoxyphenyl)methanol 97%

Supplier: Ambeed
Warning

A827362-1G A827362-250MG
77146-768EA 133.9 USD
77146-768 77146-770
(3,6-Dichloro-2-methoxyphenyl)methanol 97%
(3,6-Dichloro-2-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂
MDL Number: MFCD17171517
CAS Number: 4849-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR