1-Butyl-3-methylimidazolium acetate 99%

Supplier: Ambeed
Warning

A825016-100G A825016-25G A825016-5G
77146-746EA 157.02 USD
77146-746 77146-748 77146-750
1-Butyl-3-methylimidazolium acetate 99%
1-Butyl-3-methylimidazolium acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.27 g/mol
MDL Number: MFCD06798175
CAS Number: 284049-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR