(+)-DIOP ≥99%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (4S,5S)-(+)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (S,S)-DIOP, (((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)

A824978-1G A824978-250MG A824978-5G
77146-740EA 95.49 USD
77146-740 77146-742 77146-744
(+)-DIOP ≥99%, ee 99%
(+)-DIOP

Precaución: for Research Use Only

Formula: C₃₁H₃₂O₂P₂
MW: 498.532 g/mol
MDL Number: MFCD00009760
CAS Number: 37002-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR