[Dimethoxy(methyl)silyl]methyl methacrylate 95+% stabilised

Supplier: Ambeed
Warning

Synonyms: Methacrylic acid [Dimethoxy(methyl)silyl]methyl ester, (Dimethoxy(methyl)silyl)methyl methacrylate

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A824923-1G A824923-5G
77146-736EA 9.26 USD
77146-736 77146-738
[Dimethoxy(methyl)silyl]methyl methacrylate 95+% stabilised
[Dimethoxy(methyl)silyl]methyl methacrylate

Precaución: for Research Use Only

Formula: C₈H₁₆O₄Si
MDL Number: MFCD07777082
CAS Number: 121177-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR