4-Chloro-3-hydrazinobenzotrifluoride 95%

Supplier: Ambeed
Warning

A824762-1G A824762-250MG A824762-5G
77146-730EA 38.11 USD
77146-730 77146-732 77146-734
4-Chloro-3-hydrazinobenzotrifluoride 95%
4-Chloro-3-hydrazinobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₆ClF₃N₂
MDL Number: MFCD00052685
CAS Number: 1869-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR