Methyl 4-(bromomethyl)-3,5-difluorobenzoate 97%

Supplier: Ambeed
Danger

A824159-1G A824159-250MG
77146-720EA 221.45 USD
77146-720 77146-722
Methyl 4-(bromomethyl)-3,5-difluorobenzoate 97%
Methyl 4-(bromomethyl)-3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MDL Number: MFCD28148155
CAS Number: 1446516-72-8
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR