(2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-trans-4-F-Pro-OH·HCl
A823883-1G
A823883-250MG
A823883-5G
77146-716EA
60.48
USD
77146-716
77146-718
77747-574
(2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride 97%
(2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₉ClFNO₂ MW: 169.58 g/mol |
MDL Number:
MFCD11053638 CAS Number: 60604-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...