Chloroacetaldehyde diethyl acetal 98%

Supplier: Ambeed
Danger

Synonyms: 1-Chloro-2,2-diethoxyethane, 2-Chloro-1,1-diethoxyethane

A818566-25G A818566-100G
77146-698EA 6.37 USD
77146-698 77686-844
Chloroacetaldehyde diethyl acetal 98%
Chloroacetaldehyde diethyl acetal

Precaución: for Research Use Only

Formula: C₆H₁₃ClO₂
MW: 152.62 g/mol
MDL Number: MFCD00000949
CAS Number: 621-62-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR