(2H,4H-Spiro[benzo[b][1,4]dioxepine-3,1'-cyclopropan]-6-yl)boronic acid 97%

Supplier: Ambeed
Danger

A815625-100MG A815625-1G A815625-250MG
77146-692EA 36.08 USD
77146-692 77146-694 77146-696
(2H,4H-Spiro[benzo[b][1,4]dioxepine-3,1'-cyclopropan]-6-yl)boronic acid 97%
(2H,4H-Spiro[benzo[b][1,4]dioxepine-3,1'-cyclopropan]-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₄
MW: 220.02 g/mol
MDL Number: MFCD20441817
CAS Number: 2096335-53-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR