tert-Butyl 5-chloro-3-iodo-1H-indole-1-carboxylate 97%

Supplier: Ambeed
Warning

A814411-1G A814411-5G
77146-688EA 126.25 USD
77146-688 77146-690
tert-Butyl 5-chloro-3-iodo-1H-indole-1-carboxylate 97%
tert-Butyl 5-chloro-3-iodo-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃ClINO₂
MW: 377.60 g/mol
MDL Number: MFCD14635692
CAS Number: 1048039-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR