Methyl 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A811682-1G A811682-5G
77146-666EA 71.07 USD
77146-666 77146-668
Methyl 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride 97%
Methyl 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₃
MW: 243.68 g/mol
MDL Number: MFCD03001751
CAS Number: 672310-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR