3-(Piperidin-2-yl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: (±) Anabasine, 2-Pyridin-3-ylpiperidine

A808149-1G A808149-250MG A808149-100MG
77146-642EA 458.31 USD
77146-642 77146-644 77686-838
3-(Piperidin-2-yl)pyridine 98%
3-(Piperidin-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MW: 162.236 g/mol
MDL Number: MFCD00006370
CAS Number: 13078-04-1
UN: 3140
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR