Rubidium iodide 99.8% (metals basis)

Supplier: Ambeed
Warning

A806934-100G A806934-1G A806934-5G
77146-634EA 516.03 USD
77146-634 77146-636 77146-638
Rubidium iodide 99.8% (metals basis)
Rubidium iodide

Precaución: for Research Use Only

Formula: IRb
MDL Number: MFCD00011192
CAS Number: 7790-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR