Sodium diatrizoate 95%

Supplier: Ambeed
Danger

A806610-100MG A806610-1MG
77146-630EA 6.18 USD
77146-630 77686-830
Sodium diatrizoate 95%
Sodium diatrizoate

Precaución: for Research Use Only

Formula: C₁₁H₈I₃N₂NaO₄
MDL Number: MFCD00051544
CAS Number: 737-31-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR