(R,R)-(+)-2,3-Bis(diphenylphosphino)butane ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-Butane-2,3-diylbis(diphenylphosphine), (2R,3R)-(+)-Bis(diphenylphosphino)butane

A806486-100MG A806486-1G A806486-250MG A806486-5G
77146-626EA 12.36 USD
77146-626 77146-628 77686-828 77686-829
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane ≥98%, ee 99%
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane

Precaución: for Research Use Only

Formula: C₂₈H₂₈P₂
MDL Number: MFCD00075403
CAS Number: 74839-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR