4-tert-Butyldimethylsiloxy-1-butanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyldimethylsilyl)oxy-1-butanol, 4-((tert-Butyldimethylsilyl)oxy)butan-1-ol

A805412-100G A805412-10G A805412-1G A805412-25G A805412-5G
77146-612EA 270.53 USD
77146-612 77146-614 77146-616 77146-618 77146-620
4-tert-Butyldimethylsiloxy-1-butanol 97%
4-tert-Butyldimethylsiloxy-1-butanol

Precaución: for Research Use Only

Formula: C₁₀H₂₄O₂Si
MW: 204.38 g/mol
MDL Number: MFCD04974070
CAS Number: 87184-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR