7-Bromo-1-methyl-1H-imidazo[4,5-c]pyridin-4(5H)-one 95%

Supplier: Ambeed
Warning

A804198-100MG A804198-1G A804198-250MG
77146-606EA 23.34 USD
77146-606 77146-608 77146-610
7-Bromo-1-methyl-1H-imidazo[4,5-c]pyridin-4(5H)-one 95%
7-Bromo-1-methyl-1H-imidazo[4,5-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₆BrN₃O
MW: 228.04 g/mol
MDL Number: MFCD00182363
CAS Number: 163452-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR