1-Isopropyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A801354-100MG A801354-1G A801354-250MG A801354-5G
77146-582EA 14.42 USD
77146-582 77146-584 77146-586 77146-588
1-Isopropyl-1H-pyrazol-3-amine 95%
1-Isopropyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MDL Number: MFCD06805167
CAS Number: 857267-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR