3,4,5-Trifluorobenzamide 97%

Supplier: Ambeed
Warning

A798618-1G A798618-25G
77146-568EA 23.34 USD
77146-568 77146-570
3,4,5-Trifluorobenzamide 97%
3,4,5-Trifluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
MDL Number: MFCD00083524
CAS Number: 78324-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR