Ethyl 6-chloro-3-fluoropicolinate 97%

Supplier: Ambeed
Warning

A798317-100MG A798317-1G A798317-250MG
77146-562EA 37.07 USD
77146-562 77146-564 77146-566
Ethyl 6-chloro-3-fluoropicolinate 97%
Ethyl 6-chloro-3-fluoropicolinate

Precaución: for Research Use Only

Formula: C₈H₇ClFNO₂
MW: 203.59 g/mol
MDL Number: MFCD14698362
CAS Number: 1214363-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR