(S)-(-)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphine), (S)-MeO-BIPHEP

A797242-100MG A797242-250MG A797242-50MG A797242-1G A797242-5G
77146-556EA 13.8 USD
77146-556 77146-558 77146-560 77569-492 77618-222
(S)-(-)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 97%
(S)-(-)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

Precaución: For research use only.

Formula: C₃₈H₃₂O₂P₂
MW: 582.6070 g/mol
MDL Number: MFCD03095430
CAS Number: 133545-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR