2,9-Bis(4-chlorophenethyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 97%

Supplier: Ambeed
Warning

A795932-100MG A795932-1G A795932-250MG
77146-544EA 67.9 USD
77146-544 77146-546 77146-548
2,9-Bis(4-chlorophenethyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 97%
2,9-Bis(4-chlorophenethyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

Precaución: for Research Use Only

Formula: C₄₀H₂₄Cl₂N₂O₄
MW: 667.54 g/mol
MDL Number: MFCD23704417
CAS Number: 215726-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR