6-Bromo-5-fluoro-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%

Supplier: Ambeed
Warning

A795824-100MG A795824-250MG
77146-540EA 186.72 USD
77146-540 77146-542
6-Bromo-5-fluoro-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%
6-Bromo-5-fluoro-3,4-dihydro-2H-benzo[b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
MW: 232.04 g/mol
MDL Number: MFCD28405313
CAS Number: 1256256-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR