(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,3-difluoro-4-methoxyphenylboronic acid

A794679-1G A794679-250MG A794679-5G
77146-534EA 157.02 USD
77146-534 77146-536 77146-538
(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid 97%
(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrF₂O₃
MW: 266.83 g/mol
MDL Number: MFCD18914514
CAS Number: 2096337-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR