Carbidopa monohydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic Acid
A788203-1G
A788203-250MG
A788203-25G
A788203-25MG
A788203-5G
A788203-100G
77146-498EA
135.8
USD
77146-498
77146-500
77146-502
77146-504
77146-506
77146-508
77747-533
Carbidopa monohydrate 97%
Carbidopa monohydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂O₄ MW: 226.23 g/mol |
MDL Number:
MFCD00889211 CAS Number: 38821-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...