5-Hydroxy-2-(hydroxymethyl)-1-phenylpyridin-4(1H)-one 95%

Supplier: Ambeed
Warning

A787343-100MG A787343-1G A787343-250MG
77146-488EA 81.69 USD
77146-488 77146-490 77146-492
5-Hydroxy-2-(hydroxymethyl)-1-phenylpyridin-4(1H)-one 95%
5-Hydroxy-2-(hydroxymethyl)-1-phenylpyridin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
MDL Number: MFCD27991853
CAS Number: 70033-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR