4-Bromo-2-methylbenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A786204-1G A786204-25G A786204-5G
77146-478EA 11.33 USD
77146-478 77146-480 77146-482
4-Bromo-2-methylbenzenesulfonyl chloride 98%
4-Bromo-2-methylbenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MDL Number: MFCD00219927
CAS Number: 139937-37-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR