(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 98%

Supplier: Ambeed
Warning

A785803-1G A785803-25G A785803-5G
77146-472EA 8.24 USD
77146-472 77146-474 77146-476
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 98%
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂S₃
MDL Number: MFCD00129970
CAS Number: 53723-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR