2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-(methoxycarbonyl)phenylboronic acid pinacol ester, Methyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A783952-100MG A783952-1G A783952-250MG
77146-460EA 20.6 USD
77146-460 77146-462 77146-464
2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.10 g/mol
MDL Number: MFCD16996311
CAS Number: 1638847-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR