2-(2-Chloro-3-nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A781283-100MG A781283-1G A781283-250MG A781283-50MG
77146-452EA 172.01 USD
77146-452 77146-454 77146-456 77146-458
2-(2-Chloro-3-nitrophenyl)acetic acid 97%
2-(2-Chloro-3-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
MDL Number: MFCD18393744
CAS Number: 34915-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR