α,2-Dichlorotoluene 99%

Supplier: Ambeed
Danger

Synonyms: 2-Chlorobenzyl chloride, 1-Chloro-2-(chloromethyl)benzene

A780623-100G A780623-25G A780623-500G
77146-446EA 11.67 USD
77146-446 77146-448 77146-450
α,2-Dichlorotoluene 99%
α,2-Dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₆Cl₂
MW: 161.03 g/mol
MDL Number: MFCD00000893
CAS Number: 611-19-8
UN: 2235
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR