Dibenzo[b,d]furan-1-ol 97%

Supplier: Ambeed
Warning

A778719-250MG A778719-5G
77146-428EA 31.83 USD
77146-428 77146-430
Dibenzo[b,d]furan-1-ol 97%
Dibenzo[b,d]furan-1-ol

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MW: 184.19 g/mol
MDL Number: MFCD00463251
CAS Number: 33483-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR