3,5-Bis(bromomethyl)pyridine hydrobromide 97%
Supplier: Ambeed
|
Danger
|
A778684-250MG
A778684-1G
77146-426EA
84.88
USD
77146-426
77573-504
3,5-Bis(bromomethyl)pyridine hydrobromide 97%
3,5-Bis(bromomethyl)pyridine hydrobromide
Precaución: for Research Use Only
Formula:
C₇H₈Br₃N MW: 345.85 g/mol |
MDL Number:
MFCD27996064 CAS Number: 1118754-56-5 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...