(3,5-Dimethyl-4-isoxazolyl)methanol 96%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-(hydroxymethyl)isoxazole

A775751-100MG A775751-1MG
77146-400EA 23.34 USD
77146-400 77146-402
(3,5-Dimethyl-4-isoxazolyl)methanol 96%
(3,5-Dimethyl-4-isoxazolyl)methanol

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.1400 g/mol
MDL Number: MFCD00464226
CAS Number: 19788-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR