2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A772581-100MG A772581-1G A772581-250MG
77146-378EA 45.62 USD
77146-378 77146-380 77146-382
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-5-carbaldehyde 95%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₇BO₃S
MW: 288.17 g/mol
MDL Number: MFCD07368240
CAS Number: 953410-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR