3-(Fluoromethyl)azetidine hydrochloride 98%

Supplier: Ambeed
Warning

A770971-5G A770971-1G A770971-250MG
77146-376EA 2503.73 USD
77146-376 77569-462 77618-114
3-(Fluoromethyl)azetidine hydrochloride 98%
3-(Fluoromethyl)azetidine hydrochloride

Precaución: For research use only.

Formula: C₄H₈FN·ClH
MW: 125.572 g/mol
MDL Number: MFCD27991715
CAS Number: 1642298-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR