5-(2-Methylphenyl)-1H-tetrazole 98%

Supplier: Ambeed
Danger

Synonyms: 5-(o-Tolyl)tetrazole

A770111-250MG A770111-1G
77146-372EA 12.74 USD
77146-372 77618-112
5-(2-Methylphenyl)-1H-tetrazole 98%
5-(2-Methylphenyl)-1H-tetrazole

Precaución: for Research Use Only

Formula: C₈H₈N₄
MW: 160.18 g/mol
MDL Number: MFCD03093095
CAS Number: 51449-86-6
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR