(RS)-1-(3,5-Difluorophenyl)ethylamine 98%

Supplier: Ambeed
Danger

Synonyms: 1-(3,5-Difluorophenyl)ethanamine, 3,5-Difluoro-alpha-methylbenzylamine

A766543-250MG A766543-5G
77146-362EA 16.98 USD
77146-362 77146-364
(RS)-1-(3,5-Difluorophenyl)ethylamine 98%
(RS)-1-(3,5-Difluorophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MW: 157.16 g/mol
MDL Number: MFCD04038286
CAS Number: 321318-29-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR