4-(Pyridin-3-yl)pyrrolidin-2-one 98%

Supplier: Ambeed
Warning

A764649-100MG A764649-1G A764649-250MG
77146-356EA 43.26 USD
77146-356 77146-358 77146-360
4-(Pyridin-3-yl)pyrrolidin-2-one 98%
4-(Pyridin-3-yl)pyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MDL Number: MFCD16653126
CAS Number: 55656-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR