(R)-(+)-4-Chloro-α-methylbenzylamine 95%

Supplier: Ambeed
Danger

Synonyms: (R)-(+)-1-(4-Chlorophenyl)ethylamine, (R)-1-(4-Chlorophenyl)ethanamine, (R)-1-(4-Chlorophenyl)ethylamine

A764021-1G A764021-250MG A764021-5G
77146-348EA 73.21 USD
77146-348 77146-350 77146-352
(R)-(+)-4-Chloro-α-methylbenzylamine 95%
(R)-(+)-4-Chloro-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
MDL Number: MFCD00671639
CAS Number: 27298-99-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR