(R)-Benzyl 2-hydroxy-3-phenylpropanoate 95%

Supplier: Ambeed
Warning

A763074-1G A763074-250MG
77146-342EA 64.89 USD
77146-342 77146-344
(R)-Benzyl 2-hydroxy-3-phenylpropanoate 95%
(R)-Benzyl 2-hydroxy-3-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MDL Number: MFCD00674062
CAS Number: 7622-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR