Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4R,5R)-Dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, Dimethyl (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylate, (-)-2,3-O-Isopropylidene-L-tartaric Acid Dimethyl Ester, (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl Ester, (2R,3R)-(-)-Dimethyl-2,3-O-isopropylidene tartrate
A763060-100G
A763060-1G
A763060-5G
A763060-25G
77146-336EA
891.16
USD
77146-336
77146-338
77146-340
77552-338
Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate 98%
Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₆ MW: 218.21 g/mol |
MDL Number:
MFCD00066218 CAS Number: 37031-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...