1-(((4-Chlorophenoxy)carbonyl)oxy)ethyl isobutyrate 95%

Supplier: Ambeed
Warning

A762362-100MG A762362-1G A762362-250MG
77146-324EA 14.42 USD
77146-324 77146-326 77146-328
1-(((4-Chlorophenoxy)carbonyl)oxy)ethyl isobutyrate 95%
1-(((4-Chlorophenoxy)carbonyl)oxy)ethyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClO₅
MDL Number: MFCD29920546
CAS Number: 1622940-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR