(7-Bromo-1-benzothiophen-2-yl)methanol 97%

Supplier: Ambeed
Warning

A762156-1G A762156-5G
77146-320EA 15.45 USD
77146-320 77146-322
(7-Bromo-1-benzothiophen-2-yl)methanol 97%
(7-Bromo-1-benzothiophen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₇BrOS
MDL Number: MFCD12032282
CAS Number: 1171926-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR