2-(Dimethylamino)-7H-purin-6-ol 95%

Supplier: Ambeed
Warning

A761924-100MG A761924-250MG
77146-316EA 116.7 USD
77146-316 77146-318
2-(Dimethylamino)-7H-purin-6-ol 95%
2-(Dimethylamino)-7H-purin-6-ol

Precaución: for Research Use Only

Formula: C₇H₉N₅O
MW: 179.17 g/mol
MDL Number: MFCD00055968
CAS Number: 1445-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR